missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqPath™ 1-Step Multiplex Master Mix

Product Code. 15671832 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
0.5 mL
10 mL
5 x 1 mL
Unit Size:
0.5mL
10mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15671832 5 x 1 mL Pack of 5
15661832 0.5 mL 0.5mL
15681832 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15671832 Supplier Applied Biosystems™ Supplier No. A28526
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

TaqPath 1-Step Multiplex Master Mix is a fast, single-tube, 4X RT-qPCR mix that provides for sensitive, reproducible detection of up to four different RNA/DNA targets in a single multiplex reaction.

TaqPath 1-Step Multiplex Master Mix is a fast, single-tube, 4X RT-qPCR mix that provides for sensitive, reproducible detection of up to four different RNA/DNA targets in a single multiplex reaction. Particularly useful for diagnostics and biopharmaceuticals with applications in viral pathogen detection and high throughput gene expression workflows, the mix includes thermostable MMLV reverse transcriptase, dNTPs, UNG, MUSTANG PURPLE dye (passive reference), and thermostable Fast DNA polymerase all in a single tube. TaqPath is manufactured under a strong ISO13485 quality management system and is designed to produce consistent product performance lot after lot.

This single-tube, 4X format facilitates easy reaction set up—just add user-supplied assay and sample (Figure 1). With reproducible performance even in the presence of inhibitors, coupled with stringent quality control to minimize lot-to-lot variation, TaqPath 1-Step Multiplex Master Mix is a superior choice for your multiplex diagnostic or bioprocess quality control testing and development needs.

Features of the TaqPath 1-Step Multiplex Master Mix include:

  • High sensitivity—to detect low-copy targets with reproducible CT results
  • Broad dynamic range—with both RNA and DNA targets
  • Optimized for multiplexing—up to four targets simultaneously with MUSTANG PURPLE reference dye
  • Tolerant of inhibitors—commonly found in clinical and biopharma samples
  • Advanced manufacturing—production and process controls ensure lot-to-lot consistency
  • Compliance Package—containing most frequently requested compliance documents, available below, commonly used to comply with internal quality systems and regulatory requirements in molecular diagnostic assay and drug development

High sensitivity

To enable reproducible detection of low-titer pathogen and transcripts, TaqPath 1-Step Multiplex Master Mix has been optimized as a higher-concentration 4X master mix that allows you to input more sample into each reaction, increasing sensitivity even in low-volume reactions. Figure 1 shows the consistency of results obtained when detecting 10 copy inputs of RNA target.

Wide dynamic range compatible with RNA and DNA

TaqPath 1-Step Multiplex Master Mix has been optimized to provide high specificity and dynamic range for use with both RNA and DNA targets. Since virology labs often test for both RNA and DNA viruses, TaqPath 1-Step Multiplex Master Mix is designed to use a single protocol to assay both types of nucleic acid. This input flexibility can help streamline the number of different workflows in your lab to improve efficiency. Figure 2 demonstrates the excellent PCR linearity across a 6-log RNA dilution series, while Figure 3 exhibits a 6-log linear dynamic range for both RNA and DNA targets.

Optimized for multiplexing

TaqPath 1-Step Multiplex Master Mix has been validated for multiplexing up to four targets simultaneously, allowing for additional targets and/or controls to be run simultaneously for efficiency or quality control purposes. Rather than including ROX dye as a passive reference, the TaqPath 1-step Multiplex Master Mix includes the longer emission wavelength MUSTANG PURPLE dye as a passive reference. This longer wavelength passive reference dye enables measurement of the JUN dye, or a similar emission wavelength dye, in the channel previously used to measure ROX dye. Figure 3 demonstrates TaqPath 1-Step Multiplex Master Mix performance in 4-plex reactions with both RNA and cDNA targets.

Inhibitor tolerant

Unlike other master mixes on the market, the unique proprietary formulation of TaqPath 1-Step RT-qPCR Master Mix allows robust performance even in the presence of PCR inhibitors typically found in biopharmaceutical, molecular diagnostics, and research applications. Optimized performance in the presence of such substances that can normally inhibit PCR, such as heparin, hematin, or EDTA, helps increase confidence when working with a variety of complex clinical or biopharma samples. TaqPath master mixes demonstrate increased tolerance of inhibitors compared to competitor mixes (Figure 4).

Manufacturing production and process controls

TaqPath 1-Step Multiplex Master Mix is manufactured under a strong quality management system that utilizes traceable quality raw materials and validated operating procedures. With established controls from purchasing through QC release, TaqPath product manufacturing is designed to produce consistent product performance lot after lot. The stringent QC is important for a variety of applications, making the master mix ideal for bioprocess quality control testing, biologics adventitious testing, molecular diagnostics, and research applications.

General Purpose Reagent manufactured under cGMP

TaqPath master mixes are general purpose reagents labeled 'For Laboratory Use' and manufactured in an ISO 13485-certified and FDA-registered facility that adheres to cGMP principles.

Compliance Package

In an effort to efficiently support your compliance needs, frequently requested compliance documents are available here:

  • Statement of Quality relating to the current good manufacturing practices (cGMP) to which TaqPath master mixes adhere
  • Detailed lot-specific Certificate of Analysis (COA) (see below in Documents & Downloads section)
  • Product Stability Statement
  • Supplier Evaluation Support
    • Completed Self Assessment
    • ISO certificates by manufacturing site
  • For EU Reach compliance-related inquiries, email LSGChemicalCompliance@thermofisher.com
  • BSE/TSE Statement
  • Change Control and Notification Registration

Not finding the compliance document you're looking for? Technical Support is here to help.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 4X
Content And Storage Contains 5 x 1 mL tubes of 4 x RT-qPCR master mix sufficient for 1000 x 20 μL reactions.

Store at -10°C to -30°C in the dark.
Detection Method Primer-probe
Format Tube
PCR Method 1-step RT-qPCR
Polymerase Fast DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) QuantStudio 6 Pro Real-Time PCR System or QuantStudio 7 Pro, QuantStudio 3 or 5, QuantStudio 6/QuantStudio 7 Flex, QuantStudio 12K Flex, QuantStudio Dx, ViiA 7, 7500 , 7500 Fast, 7500 Fast Dx
Multiplex Capability Up to 4-plex
Passive Reference Dye Mustang Purple
Product Line TaqPath
Product Type qPCR Master Mix
Quantity 5 x 1 mL
Sensitivity Down to 10 copies
Shipping Condition Dry Ice
Sufficient For 1000 Reactions of 20 μL
Target Specificity RNA only, Both RNA and DNA
For Use With (Application) Gene Expression, Microbial Detection
Thermostability Not Applicable
No. of Reactions 1000 Reactions
Sample Type DNA, RNA
Volume 5 x 1 mL
Includes dUTP, UNG/UDG
Inhibitor Tolerance (Simple) High
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.