Learn More
Applied Biosystems™ TaqPath™ Menu ∣ GeneProof™ Human Papillomavirus (HPV) Screening PCR Kit
Shop All Applied Biosystems ProductsDescription
The TaqPath Menu | GeneProof Human Papillomavirus (HPV) Screening PCR Kit is a real-time PCR testing solution that can detect 24 high-risk HPV types. The kit is approved for use on the QuantStudio 5 Real Time PCR System (96-well, 0.2 mL block) and the Applied Biosystems 7500 Real-Time PCR System (96-well).
Features of the TaqPath Menu | GeneProof Human Papillomavirus (HPV) Screening PCR Kit include:
• Differentiates high risk types 16, 18, and 45
• Minimizes contamination—the master mix contains uracil-DNA glycosylase (UNG) and dUTPs, reducing carryover contamination
• Compatible with a wide range of real-time PCR devices
Intended use
The TaqPath Menu | GeneProof Human Papillomavirus (HPV) Screening PCR Kit is a real-time PCR testing solution that provides qualitative detection of 24 high-risk HPV types. The PCR kit can be used in combination with a manual or automated extraction system. The kit is intended for diagnostics, aid to diagnosis and screening. It is designed for professional use in laboratories with trained staff.
Specifications
Specifications
| Content And Storage | • Master mix • Positive Control Store at –20 ± 5°C. |
| Control Template | Proper sampling, DNA extraction efficiency, and PCR inhibition control by endogenous Internal control (GAPDH gene) |
| PCR Method | qPCR |
| Technique | Qualitative Analysis |
| For Use With (Equipment) | QuantStudio™ 3 Real-Time PCR System, QuantStudio™ 5 Real-Time PCR System, Applied Biosystems 7500 Real-Time PCR System, AMPLilab Real-Time PCR System, AriaMx Real-Time PCR System, CFX Connect™ Real-Time PCR Detection System, CFX96™/Dx Real-Time PCR Detection System, croBEE Real-Time PCR System, Gentier 96E/96R Real-Time PCR System1, LightCycler™ 4802, LineGene 9600 Plus, Rotor-Gene Q4, SLAN™ Real-Time PCR System, Stratagene Mx3005p qPCR System |
| Label or Dye | FAM |
| Product Line | TaqPath |
| Quantity | 100 reactions kit |
| Sensitivity | Diagnostic: 100% (CI95%: 87.7–100%) |
| Target | E1/E2 gene |
| Show More |
Available in select countries only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.