missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqPath™ qPCR Master Mix, CG
GREENER_CHOICE

Product Code. 15820843 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Tests:
2500 Reactions
5000 Reactions
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
15820843 5000 Reactions Each
16268100 2500 Reactions Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15820843 Supplier Applied Biosystems™ Supplier No. A15298
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Designed and all lots are functionally tested to help ensure lot-to-lot reproducibility for Ct consistency and dynamic range across a wide variety of assays.

  • Efficient and linear detection of up to 8 logs with gene expression or miRNA assays
  • Enables reliable detection of low-copy templates with reproducible CT results
  • Robust multiplexing performance with exogenous or endogenous targets
  • Engineered to retain consistent performance in preassembled reactions for up to 48 hours
  • Demonstrates increased tolerance to inhibitors compared to competitor mixes
  • Manufactured with stringent production and process controls to help ensure lot-to-lot consistency
  • Labeled “For Laboratory Use”
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in refrigerator (2° to 8°C).
Detection Method Primer-probe
For Use With (Equipment) QuantStudio™ 12K Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6, QuantStudio™ 7
No. of Tests 5000 Reactions
Passive Reference Dye ROX (Pre-mixed)
Quantity 1 x 50mL
For Use With (Application) Real Time PCR (qPCR), Gene Expression, miRNA analysis
Sample Type DNA, cDNA

For laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.