missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid, ACS Reagent Grade, Ricca Chemical

Product Code. 31751080
Change view
Click to view available options
Quantity:
125 g
500 g
Packaging:
White Poly Wide Mouth Bottle
Unit Size:
125g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
31751080 500 g White Poly Wide Mouth Bottle 500g
31749550 125 g White Poly Wide Mouth Bottle 125g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31751080 Supplier Ricca Chemical Company Supplier No. RDCT0030500B1
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 87-69-4
Molecular Formula C4H6O6
Molecular Weight (g/mol) 150.09
MDL Number MFCD00064207
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
PubChem CID 444305
ChEBI CHEBI:15671
IUPAC Name 2,3-dihydroxybutanedioic acid
SMILES OC(C(O)C(O)=O)C(O)=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Percent Purity Min. 99%
Grade ACS
Quantity 500 g
Packaging White Poly Wide Mouth Bottle
Chemical Name or Material Tartaric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.