missing translation for 'onlineSavingsMsg'
Learn More

Tartaric Acid AR Granular, Macron Fine Chemicals™

Product Code. 15558674
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15558674 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558674 Supplier Avantor Macron Supplier No. 231204
  • ONLINE EXCLUSIVE PRICE
    234.00 €
    225.96 € / 500g
    Save 8.04 €
    3% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 87-69-4
Quantity 500 g
Packaging POLYSTORMOR
Physical Form Granular
Molecular Formula C4H6O6
MDL Number MFCD00064207
Synonym l-tartaric acid, l-+-tartaric acid, l +-tartaric acid, 2r,3r-2,3-dihydroxysuccinic acid, tartaric acid, 2r,3r-2,3-dihydroxybutanedioic acid, r,r-tartaric acid, +-l-tartaric acid, dextrotartaric acid, l-threaric acid
InChI Key FEWJPZIEWOKRBE-UHFFFAOYNA-N
SMILES OC(C(O)C(O)=O)C(O)=O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid
Molecular Weight (g/mol) 150.09
PubChem CID 444305
ChEBI CHEBI:15671
Formula Weight 150.09
Chemical Name or Material Tartaric Acid
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Tartaric Acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: L(+)-Tartaric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.