missing translation for 'onlineSavingsMsg'
Learn More

Tartronic acid, 97%

Product Code. 11450197
Change view
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450197 0.5 g 0.5g
11440197 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450197 Supplier Thermo Scientific Alfa Aesar Supplier No. B21640.02
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 80-69-3 | C3H4O5 | 120.06 g/mol

Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.

Solubility
Soluble in water and alcohol.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-69-3
Molecular Formula C3H4O5
Molecular Weight (g/mol) 120.06
MDL Number MFCD00004237
InChI Key ROBFUDYVXSDBQM-UHFFFAOYSA-N
Synonym tartronic acid, hydroxymalonic acid, 2-hydroxymalonic acid, propanedioic acid, hydroxy, tartronicacid, 2-tartronic acid, hydroxypropanedioic acid, 2-hydroxymalonate, malonic acid, hydroxy, hydroxymalonate
PubChem CID 45
ChEBI CHEBI:16513
IUPAC Name 2-hydroxypropanedioic acid
SMILES C(C(=O)O)(C(=O)O)O

Specifications

Melting Point ∼151°C (decomposition)
Quantity 0.5 g
Assay Percent Range 0.98
Beilstein 1209791
Merck Index 14,9073
Solubility Information Soluble in water and alcohol.
Formula Weight 120.06
Percent Purity 98%
Chemical Name or Material Tartronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.