missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tasimelteon

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Tasimelteon is a high affinity and selective dual melatonin receptor (MT1/MT2) agonist (Ki values are 0.304 nM and 0.0692 nM, respectively). It has no significant interaction with any other commonly screened receptors or enzyme binding sites. Tasimelteon induces a potent, concentration-dependent inhibition of forskolin-stimulated cAMP accumulation in NIH-3T3 cells stably expressing human MT1 (EC50 = 0.74 nM) or expressing human MT2 (EC50 = 0.1 nM) receptors. Tasimelteon is a circadian regulator.
Specifications
Specifications
| Chemical Name or Material | N-[[(1 R,2 R)-2-(2,3-Dihydro-4-benzofuranyl)cyclopropyl]methyl]propanamide |
| CAS | 609799-22-6 |
| Quantity | 50 mg |
| Target | Melatonin (MT) Receptor Agonists |
| Molecular Formula | C15H19NO2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction