missing translation for 'onlineSavingsMsg'
Learn More

Taurine, ≥98.5%, Molecular Biology Grade, Ultrapure

Product Code. 15835368 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15835368 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15835368 Supplier Thermo Scientific Supplier No. J75824.A1
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-35-7 | C2H7NO3S | 125.14 g/mol

 

TRUSTED_SUSTAINABILITY

Specifications

CAS 107-35-7
Melting Point >300°C
Packaging Plastic bottle
Molecular Formula C2H7NO3S
MDL Number MFCD00008197
Quantity 1 kg
Beilstein 1751215
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
SMILES NCCS(O)(=O)=O
IUPAC Name 2-aminoethanesulfonic acid
Molecular Weight (g/mol) 125.14
PubChem CID 1123
ChEBI CHEBI:15891
Formula Weight 125.15
Percent Purity ≥98.5%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Taurine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.