missing translation for 'onlineSavingsMsg'
Learn More

Taurine, 99+%, Pure, Fisher Chemical

Product Code. 10614932 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10614932 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10614932 Supplier Fisher Chemical Supplier No. T/0300/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 107-35-7
Melting Point 300°C
Color White
pH 5
Packaging Plastic powder jar
Molecular Formula C2H7NO3S
MDL Number MFCD00008197
Quantity 500 g
Synonym taurine, tauphon, ethanesulfonic acid, 2-amino, l-taurine, 2-aminoethylsulfonic acid, o-due, 2-sulfoethylamine, taufon, aminoethanesulfonic acid, aminoethylsulfonic acid
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
SMILES NCCS(O)(=O)=O
IUPAC Name 2-aminoethanesulfonic acid
Molecular Weight (g/mol) 125.14
PubChem CID 1123
ChEBI CHEBI:15891
Formula Weight 125.14g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.