missing translation for 'onlineSavingsMsg'
Learn More

Taurine, MedChemExpress

Product Code. 16480888
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16480888 5 g 5g
16422149 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16480888 Supplier Medchem Express Supplier No. HYB0351
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Taurine, a sulphur-containing amino acid and an organic osmolyte involved in cell volume regulation, provides a substrate for the formation of bile salts, and plays a role in the modulation of intracellular free calcium concentration. Taurine has the ability to activate autophagy in adipocytes.

Taurine, a sulphur-containing amino acid and an organic osmolyte involved in cell volume regulation, provides a substrate for the formation of bile salts, and plays a role in the modulation of intracellular free calcium concentration. Taurine has the ability to activate autophagy in adipocytes.

Specifications

Chemical Name or Material Taurine
CAS 107-35-7
Color White
Purity Grade Notes Research
Molecular Formula C2H7NO3S
Quantity 5 g
Synonym 2-Aminoethanesulfonic acid
Solubility Information H2O : 25 mg/mL (199.76 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (7.99 mM; Need ultrasonic)
SMILES NCCS(=O)(O)=O
Molecular Weight (g/mol) 125.15
Formula Weight 125.15
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.