missing translation for 'onlineSavingsMsg'
Learn More

Taurine, TRC

Product Code. 17919434
Change view
Click to view available options
Quantity:
10 mg
1 g
10 g
Unit Size:
10g
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17919434 10 mg 10mg
17918984 10 g 10g
17917624 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17919434 Supplier Toronto Research Chemicals Supplier No. TRCT00785010MG
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2 H7 N O3 S
Molecular Weight (g/mol) 125.15
Synonym 2-Aminoethanesulfonic Acid, Taurine (8CI), 2-Aminoethanesulfonic acid, 1-Aminoethane-2-sulfonic acid, 2-Aminoethane-1-sulfonic acid, 2-Aminoethylsulfonic acid, 2-Sulfoethylamine, NSC 32428, O-Due, Taufon, Taukard, Tauphon, Tauramin, β-Aminoethylsulfonic acid
IUPAC Name 2-aminoethanesulfonic acid
SMILES NCCS(=O)(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C2H7NO3S/c3-1-2-7(4,5)6/h1-3H2,(H,4,5,6)
Formula Weight 125.0147
Percent Purity >95%
Chemical Name or Material Taurine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.