missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Taurocholic acid, sodium salt hydrate, 98%

Product Code. 10701701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10701701 1 g Glass bottle 1g
10617012 5 g Glass bottle 5g
10629453 25 g Glass bottle 25g
11995541 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10701701 Supplier Thermo Scientific Chemicals Supplier No. 230370010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

Taurocholic acid, sodium salt hydrate, CAS # 312693-83-7, also known as sodium taurocholate hydrate, is a bile acid taurine conjugate form in the liver with detergent capabilities. | CAS: 345909-26-4 | C26H44NNaO7S | 537.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Taurocholic acid, sodium salt hydrate is an anionic bile acid detergent synthesized from cholic acid
  • This compound shows a choleretic activity promoting the secretion of bile from the liver

Applications

  • Taurocholic acid is used in the emulsification of lipids/ fats
  • It is used for protein solubilization and in the preparation of digestive enzymes for research applications
  • It can be used to determine conjugated and unconjugated bile salts in serum and jejunal fluid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 345909-26-4
Molecular Formula C26H44NNaO7S
Molecular Weight (g/mol) 537.69
MDL Number MFCD00150819
InChI Key JAJWGJBVLPIOOH-IZYKLYLVSA-M
Synonym Sodium taurocholate hydrate
PubChem CID 23687511
SMILES [Na+].[H][C@@]12CC[C@H]([C@H](C)CCC(=O)NCCS([O-])(=O)=O)[C@@]1(C)[C@@H](O)C[C@@]1([H])[C@@]2([H])[C@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C

Specifications

Color Undesignated
Physical Form Powder
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
Specific Rotation +20.0° to +25.8° (20°C, 589nm) (c=3, water)
Water 7% max.
IUPAC Name sodium 2-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]ethane-1-sulfonate
Formula Weight 537.7
Percent Purity ≥97.5% (TLC)
Chemical Name or Material Taurocholic acid, sodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.