missing translation for 'onlineSavingsMsg'
Learn More

Taurolithocholic Acid Sodium Salt, TRC

Product Code. 30004246
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004246 10 mg 10mg
30010490 5 mg 5mg
30003982 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004246 Supplier Toronto Research Chemicals Supplier No. TRCT00910010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6042-32-6
Molecular Formula C26 H44 N O5 S . Na
Molecular Weight (g/mol) 505.69
Synonym Ethanesulfonic acid, 2-[[(3α,5β)-3-hydroxy-24-oxocholan-24-yl]amino]-, sodium salt (1:1), Ethanesulfonic acid, 2-[[(3α,5β)-3-hydroxy-24-oxocholan-24-yl]amino]-, monosodium salt (9CI), Taurine, N-(3α-hydroxy-5β-cholan-24-oyl)-, monosodium salt (8CI), Taurolithocholic acid, sodium salt (6CI,7CI), NSC 681057, Sodium taurolithocholate, Taurolithocholic acid sodium salt, 2-[[(3α,5β)-3-Hydroxy-24-oxocholan-24-yl]amino]ethanesulfonic acid sodium salt, 3α-Hydroxy-5β-cholanoyltaurine sodium
IUPAC Name sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,13R,14S,17R)-3-hydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate
SMILES [Na+].C[C@H](CCC(=O)NCCS(=O)(=O)[O-])[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C26H45NO5S.Na/c1-17(4-9-24(29)27-14-15-33(30,31)32)21-7-8-22-20-6-5-18-16-19(28)10-12-25(18,2)23(20)11-13-26(21,22)3;/h17-23,28H,4-16H2,1-3H3,(H,27,29)(H,30,31,32);/q;+1/p-1/t17-,18-,19-,20+,21-,22+,23+,25+,26-;/m1./s1
Formula Weight 505.2838
Chemical Name or Material Taurolithocholic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.