missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Taxol

Product Code. 30001192
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001192 10 mg 10mg
18732805 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001192 Supplier R&D Systems™ Supplier No. 1097/10
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Promotes assembly and inhibits disassembly of microtubules

Taxol, also known as paclitaxel, promotes and stabilizes tubulin polymerization, causing cell cycle arrest. In CCRF-HSB-2 cells, Taxol induces autocatalytic activation of caspase-10, triggering apoptosis. Taxol is an antitumor agent. In a mouse model of bladder cancer Taxol decreases tumor growth. Taxol also prolongs survival of mice transplanted with human ovarian carcinoma xenografts. Taxol improves neurological outcome and magnetic resonance imaging biomarkers after traumatic brain injury in mice.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33069-62-4
Molecular Formula C47H51NO14

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 853.92
Purity >99%
Molecular Weight (g/mol) 853.92
Chemical Name or Material Taxol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.