missing translation for 'onlineSavingsMsg'
Learn More

Tazobactam sodium

Product Code. 15457499
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457499 100 mg 100mg
15447499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457499 Supplier Thermo Scientific Alfa Aesar Supplier No. J66226.MC
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases. | CAS: 89785-84-2 | C10H12N4NaO5S | 323.279 g/mol

Tazobactam sodium is a beta-Lactamase inhibitor, which is used with penicillinase inhibition studies and beta-lactam antibiotics to enhance their effect. It serves as an antibacterial penicillin derivative, which inhibits the action of bacterial beta-lactamases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89785-84-2
Molecular Formula C10H12N4NaO5S
Molecular Weight (g/mol) 323.279
MDL Number MFCD00917472
InChI Key ZUNWFYFXHGWFRA-QVUDESDKSA-N
Synonym tazobactam sodium salt
PubChem CID 66636300
IUPAC Name (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-(triazol-1-ylmethyl)-4$l^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
SMILES CC1(C(N2C(S1(=O)=O)CC2=O)C(=O)O)CN3C=CN=N3.[Na]

Specifications

Melting Point 140°C to 147°C
Boiling Point 77°C
Flash Point >110°C (230°F)
Packaging Glass bottle
Quantity 100 mg
Solubility Information Soluble in water.Soluble in water and methanol.
Formula Weight 322.27
Physical Form Powder
Chemical Name or Material Tazobactam sodium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.