missing translation for 'onlineSavingsMsg'
Learn More

Tazobactam Sodium Salt, TRC

Product Code. 30426605
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426605 50 mg 50mg
30411729 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30426605 Supplier Toronto Research Chemicals Supplier No. TRCT01010050MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tazobactam Sodium Salt
CAS 89785-84-2
Purity Grade Notes HPLC
Molecular Formula C10 H11 N4 O5 S . Na
Quantity 50 mg
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','3-methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-','4,4-dioxide','sodium salt (1:1)','(2S,3S,5R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','3-methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-','4,4-dioxide','sodium salt','(2S,3S,5R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid','3-methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-','4,4-dioxide','sodium salt','[2S-(2α,3β,5α)]-, 1H-1,2,3-Triazole','4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid deriv., CL 307579, Tazobactam sodium, Tazobactam sodium salt, YTR 830, (2S,3S,5R)-3-Methyl-7-oxo-3-(1H-1,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 4,4-dioxide sodium salt
InChI Formula InChI=1S/C10H12N4O5S.Na/c1-10(5-13-3-2-11-12-13)8(9(16)17)14-6(15)4-7(14)20(10,18)19;/h2-3,7-8H,4-5H2,1H3,(H,16,17);/q;+1/p-1/t7-,8+,10+;/m1./s1
SMILES [Na+].C[C@]1(Cn2ccnn2)[C@@H](N3[C@@H](CC3=O)S1(=O)=O)C(=O)[O-]
IUPAC Name sodium;(2S,3S,5R)-3-methyl-4,4,7-trioxo-3-(triazol-1-ylmethyl)-4λ^{6}-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 322.27
Formula Weight 322.0348
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.