missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TBB

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
A cell-permeable, selective inhibitor of casein kinase-2 (CK2) (IC50 = 0.9 and 1.6 μM for rat liver and human recombinant CK2 respectively). Exhibits modest discrimination between CK2 subunits, with Ki values ranging from 80 nM to 210 nM. Acts in an ATP/GTP-competitive manner and displays one to two orders of magnitude selectivity over a panel of 33 protein kinases.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 434.71 |
| Purity | 98% |
| Molecular Weight (g/mol) | 434.711 |
| Chemical Name or Material | TBB |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction