missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TBTA

Product Code. 30000038
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30000038 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30000038 Supplier Tocris Bioscience™ Supplier No. 7862/100
  • ONLINE EXCLUSIVE PRICE
    137.00 €
    129.15 € / 100mg
    Save 7.85 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Click chemistry auxiliary reagent; stabilizing ligand for copper-catalyzed azide-alkyne cycloaddition reaction

TBTA is a click chemistry auxiliary reagent. It is a stabilizing ligand for copper-catalyzed azide-alkyne cycloaddition reactions.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
CAS 510758-28-8
Quantity 100 mg
Target General Laboratory Research Reagent
Molecular Formula C30H30N10
Purity 0.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.