missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC ASK 10
Description
Potent ASK1 inhibitor (IC50 = 14 nM); displays selectivity for ASK1 over other kinases including ASK2 (IC50 = 0.51 μM), MEKK1, TAK1, IKKβ, ERK1, JNK1, p38α, GSK-3β, PKCθ and B-raf (IC50 values are > 10 μM). Blocks downstream JNK1/p38 phosphorylation in cells. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 432.35 |
| Purity | >98% |
| Molecular Weight (g/mol) | 432.349 |
| Chemical Name or Material | TC ASK 10 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?