missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC ASK 10
GREENER_CHOICE

Product Code. p-4817041
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001618

Brand: Tocris Bioscience™ 4825/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective ASK1 inhibitor; orally bioavailable

Potent ASK1 inhibitor (IC50 = 14 nM); displays selectivity for ASK1 over other kinases including ASK2 (IC50 = 0.51 μM), MEKK1, TAK1, IKKβ, ERK1, JNK1, p38α, GSK-3β, PKCθ and B-raf (IC50 values are > 10 μM). Blocks downstream JNK1/p38 phosphorylation in cells. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1005775-56-3
Molecular Formula C21H23Cl2N5O

Specifications

Quantity 10 mg
Formula Weight 432.35
Purity >98%
Molecular Weight (g/mol) 432.349
Chemical Name or Material TC ASK 10
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.