missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC NTR1 17

Description
TC NTR1 17 is a non-peptide neurotensin receptor 1 (NTS1) partial agonist (EC50 <15.6 nM and Emax = 63% of neurotensin response in Ca2+ mobilization assay). Exhibits over 50-fold selectivity for NTS1 over NTS2 and GPR35.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | TC NTR1 17 |
| Purity | >99% |
| Molecular Weight (g/mol) | 522.98 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?