Learn More
Tocris Bioscience™ TC-S 7001
GREENER_CHOICE
Potent and highly selective ROCK inhibitor; orally active
Brand:  Tocris Bioscience™ 4961/1
Encompass
Product Code. 15736356
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
TC-S 7001 (also known as BAY-549) is a potent and highly selective ROCK inhibitor (IC50 values are 0.6 and 1.1 nM for human ROCK1 and ROCK2, respectively), which exhibits >200-fold selectivity over TRK and FLT3 receptors, and >900-fold selectivity over a panel of other kinases and cardiovascular relevent enzymes and receptors. TC-S 7001 reduces blood pressure in normotensive and hypertensive rats, and is orally active.
Specifications
Specifications
6-Chloro-N4-[3,5-difluoro-4-[(3-methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)oxy]phenyl]-2,4-pyrimidinediamine
867017-68-3
Azaindole-1