missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC-S 7001

Description
TC-S 7001 (also known as BAY-549) is a potent and highly selective ROCK inhibitor (IC50 values are 0.6 and 1.1 nM for human ROCK1 and ROCK2, respectively), which exhibits >200-fold selectivity over TRK and FLT3 receptors, and >900-fold selectivity over a panel of other kinases and cardiovascular relevent enzymes and receptors. TC-S 7001 reduces blood pressure in normotensive and hypertensive rats, and is orally active.
Specifications
Specifications
| Chemical Name or Material | 6-Chloro-N4-[3,5-difluoro-4-[(3-methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)oxy]phenyl]-2,4-pyrimidinediamine |
| CAS | 867017-68-3 |
| Synonym | Azaindole-1 |
| Quantity | 1 mg |
| Target | Rho-kinase Inhibitors |
| Molecular Formula | C18H13ClF2N6O |
| Formula Weight | 402.79 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?