missing translation for 'onlineSavingsMsg'
Learn More

HUBER TC50-F Immersion Chiller

Product Code. 10003223
Change view
Click to view available options
Model:
TC50-F
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
10003223 TC50-F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10003223 Supplier HUBER Supplier No. 3004.0020.00
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For many applications immersion coolers are also an environmentally friendly alternative to mains water cooling. The TC immersion coolers are suitable for unsupervised use, for cooling tasks which require continuous cooling or as a variant with temperature control and a Pt100 probe.

  • TC50-F Immersion cooler, with flexible evaporator coil and natural refrigerant
  • Rapid cooling of fluids
  • Ideal for counter-cooling or as a replacement of dry ice
  • LED temperature display
  • Easy eration with only three keys
  • Compact, space-saving construction
  • Robust housing with carrying grips
  • High quality stainless steel immersion probe
  • Temperature range: -50°C to 50°C
  • Cooling apacity (0°C): 0.3kW

Specifications

Temperature Range -50°C to +50°C
Model TC50-F
Voltage 208/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.