missing translation for 'onlineSavingsMsg'
Learn More

TCEP Hydrochloride, TRC

Product Code. 30004865
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004865 10 g 10g
30009478 1 g 1g
30003306 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004865 Supplier Toronto Research Chemicals Supplier No. TRCT01250010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51805-45-9
Formula Weight 286.0373
InChI Formula InChI=1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H
IUPAC Name 3-[bis(2-carboxyethyl)phosphanyl]propanoic acid;hydrochloride
Molecular Formula C9 H15 O6 P . Cl H
Molecular Weight (g/mol) 286.65
Percent Purity >95%
SMILES Cl.OC(=O)CCP(CCC(=O)O)CCC(=O)O
Synonym Tris (2-Carboxyethyl) Phosphine Hydrochloride (TCEP)
Chemical Name or Material TCEP Hydrochloride
Grade HPLC
Show More Show Less

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.