missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TCID
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TCID is a selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor (IC50 = 0.6 μM). Exhibits >100-fold selectivity for UCH-L3 over UCH-L1. Diminishes glycine transporter GlyT2 ubiquitination in brain stem and spinal cord primary neurons.
Specifications
Specifications
| Purity | >99% |
| Quantity | 50 mg |
| Product Type | TCID |
| Molecular Weight (g/mol) | 283.92 |
| Inhibitors | TCID |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction