missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TCID

Product Code. p-5546063
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18771558

Brand: Tocris Bioscience™ 5179/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor

TCID is a selective ubiquitin C-terminal hydrolase-L3 (UCH-L3) inhibitor (IC50 = 0.6 μM). Exhibits >100-fold selectivity for UCH-L3 over UCH-L1. Diminishes glycine transporter GlyT2 ubiquitination in brain stem and spinal cord primary neurons.
TRUSTED_SUSTAINABILITY

Specifications

Purity >99%
Quantity 10 mg
Product Type TCID
Molecular Weight (g/mol) 283.92
Inhibitors TCID

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.