missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCTN1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
196.00 € - 468.00 €
Specifications
| Antigen | TCTN1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TCTN1 Polyclonal antibody specifically detects TCTN1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| TCTN1 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience, Stem Cells | |
| PBS (pH 7.3), 50% glycerol | |
| 79600 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| TECT1FLJ21127, tectonic 1, tectonic family member 1, tectonic-1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-310 of human TCTN1 (NP_001076006.1). NFTANPPQRVFELVDQINPSIFCIHITNYKPALSFINPEVPDENNFDTLMKTSDGFTLNAESYVSFTTKLDIPTAAKYEYGVPLQTSDSFLRFPSSLTSSLCTDNNPAAFLVNQAVKCTRKINLEQCEEIEALSMAFYSSPEILRVPDSRKKVPITVQSIVIQSLNKTLTRREDTDVLQPT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title