missing translation for 'onlineSavingsMsg'
Learn More

TCV-309 chloride, MedChemExpress

Product Code. 16493687
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493687 50 mg 50mg
16491769 1 mg 1mg
16480158 5 mg 5mg
16467698 10 mg 10mg
16407586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493687 Supplier Medchem Express Supplier No. HY19121A
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TCV-309 chloride is a potent and specific platelet activating factor (PAF) antagonist. TCV-309 chloride specifically inhibits PAF-induced aggregation of rabbit and human platelets, and [3H]PAF binding to rabbit platelet microsomes with IC50 values of 33 nM, 58 nM and 27 nM, respectively. TCV-309 chloride has beneficial effects in anaphylactic shock.

TCV-309 chloride is a potent and specific platelet activating factor (PAF) antagonist. TCV-309 chloride specifically inhibits PAF-induced aggregation of rabbit and human platelets, and [3H]PAF binding to rabbit platelet microsomes with IC50 values of 33 nM, 58 nM and 27 nM, respectively. TCV-309 chloride has beneficial effects in anaphylactic shock.

Specifications

Chemical Name or Material TCV-309 chloride
CAS 121494-09-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H34BrClN4O4
Quantity 50 mg
Solubility Information DMSO : ≥ 6.3 mg/mL (10.00 mM)
SMILES CCC[N+]1=CC(C(N(CCC(NCCOC(N2CC3=C(C=CC=C3)CC2)=O)=O)C4=CC=CC=C4)=O)=CC(Br)=C1.[Cl-]
Molecular Weight (g/mol) 629.97
Formula Weight 629.97
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.