missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ tDHU, acid

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
tDHU, acid is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a benzoic acid functional handle for ready conjugation to a linker/target protein ligand.
Part of a range of functionalized tool molecules for PROTAC® Degrader R&D.
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-3-(tetrahydro-2,4-dioxo-1(2 H)-pyrimidinyl)benzoic acid |
| CAS | 2377643-37-1 |
| Quantity | 50 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C12H12N2O4 |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction