Learn More
Tocris Bioscience™ TDZD 8
Selective non-ATP competitive inhibitor of GSK 3β
Brand:  Tocris Bioscience™ 6092/50
Encompass
Product Code. 15747216
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
TDZD 8 is a selective non-ATP competitive inhibitor of GSK 3β (IC50 = 2 μM); thiadiazolidinone deriviative. Does not inhibit Cdk-1/cyclin B, CK-II, PKA or PKC at >100 μM. Reduces severity of L-dopa-induced dyskinesia in a Parkinson's disease in vivo model.
Specifications
Specifications
White
327036-89-5
222.26g/mol
Soluble to 100mM in DMSO, 100mM in ethanol
Inhibits GSK 3β
Inhibitor
C10H10N2O2S
NP 01139
Store at -20°C