missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Teats for Pasteur pipettes
Description
For use with Soda-lime Pasteur pipettes.
- For use with Soda-lime Pasteur pipettes
Specifications
Specifications
| Description | Teats for Pasteur pipettes |
| Color | Clear |
| Material | Silicone |
| For Use With (Equipment) | Soda-lime Pasteur pipettes |
| Quantity | 100/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?