missing translation for 'onlineSavingsMsg'
Learn More

Teats for Pasteur pipettes

Product Code. 15758007
Change view
Click to view available options
Color:
Clear
Red
Unit Size:
Pack of 100
Product Code. Color unitSize
15758007 Red Pack of 100
15748007 Clear Pack of 100
2 options
This item is not returnable. View return policy

Product Code. 15758007

Brand: MBL BPP004

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Soda-lime Pasteur pipettes.

  • For use with Soda-lime Pasteur pipettes
TRUSTED_SUSTAINABILITY

Specifications

Description Teats for Pasteur pipettes
Color Red
For Use With Soda Lime Pasteur Pipette
Material Natural Latex Rubber
For Use With (Equipment) Soda-lime Pasteur pipettes
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.