Learn More
Medline™ Technico™ DIGIT-090 Pocket Refractometer
Product Code. 10313295
Change view
Click to view available options
Scale Range:
0 to 42 Brix, 42 to 71 Brix, 71 to 90 Brix
Unit Size:
Each
Product Code. Scale Range Unit Size
10313295 0 to 42 Brix, 42 to 71 Brix, 71 to 90 Brix Each
1 options
Product Code. 10313295 Supplier Medline™ Supplier No. 8100.0900M
Estimated Shipment: 04-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Type Pocket Refractometer
Scale Range 0 to 42 Brix, 42 to 71 Brix, 71 to 90 Brix
Model DIGIT-090
Calibration Zero calibration with distilled water at 20°
Quantity 1 Each