Learn More
Juchheim Laborgeraete™ Teclu Safety Burner
A high level of stability is achieved due to the high weight and low center of gravity of the burners.
Brand:  Juchheim Laborgeraete™ 01262 DE
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10031262
  • 50.80 € / Each
Estimated Shipment: 28-04-2023
to see stock.
Description
Description
  • For operation with option of natural gas or propane gas
  • With needle valve
  • With DVGW certification
  • With air control screen
  • Particularly good stand due to heavy, high-quality material
Specifications
Specifications
Bunsen Burner
Brass
Needle Valve
Natural Gas
Gas consumption approx. 116 L/h (0.0813 kg/h), Heating capacity approx. 3398 kJ/h