Learn More
Juchheim Laborgeraete™ Teclu Safety Burner
A high level of stability is achieved due to the high weight and low center of gravity of the burners.
55.55 € - 181.00 €
Product Code | Brand | Gas Type | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Gas Type | Includes | Price | Quantity & Availability | ||||
10103152
|
Juchheim Laborgeraete™
01374 DP |
Propane | Needle Valve, Safety Valve, and Automatic Gas shut-off |
181.00 €
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
10091292
|
Juchheim Laborgeraete™
01374 DE |
Natural Gas | Needle Valve, Safety Valve, and Automatic Gas shut-off |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10031262
|
Juchheim Laborgeraete™
01262 DE |
Natural Gas | Needle Valve |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- For operation with option of natural gas or propane gas
- With needle valve
- With DVGW certification
- With air control screen
- Particularly good stand due to heavy, high-quality material
Specifications
Bunsen Burner | |
Brass |