Learn More
Sampling Systems™ TeleSwab™ Telescopic Swabbing Tool Complete Kits
Product Code. 15774642 Shop All Sampling Systems Products
Change view
Click to view available options
Style:
Kit 1
Kit 2
Includes:
Angle Adapter, Swab Holder
Template Holder, Swabbing Template
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Style Includes unitSize
15774642 Kit 1 Angle Adapter, Swab Holder 1 set
15491885 Kit 2 Template Holder, Swabbing Template 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15774642 Supplier Sampling Systems™ Supplier No. 8011K01
Only null left
Add to basket
Add to basket
The TeleSwab™ Telescopic Swabbing Tool allows operators to quickly and easily take swabs from previously inaccessible places.
  • Ideal for environmental swabbing and for use with large items of processing equipment
  • Extendable handle that can be extended up to 3m
  • Adjustable adapter for angling swabs to meet the contours of the item being sampled
  • PTFE template tool allows a fixed area to be swabbed
  • Two different complete kits are available
  • Kit 1 contains one telescopic pole, one angle adapter and one swab holder
  • Kit 2 contains one telescopic pole, one template holder and one swabbing template with a hole cut to customer’s requirements
Specifications
Product Type Swabbing Kit
Includes Angle Adapter, Swab Holder
Handle Type Telescoping
Style Kit 1
For Use With (Application) Environmental swabbing
Length (Metric) Handle Extendable to 3 m