missing translation for 'onlineSavingsMsg'
Learn More

Tellurium, 99.8%, (trace metal basis), powder, -200 mesh

Product Code. 10716443 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10716443 25 g Glass bottle 25g
10676871 100 g Glass bottle 100g
10005973 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10716443 Supplier Thermo Scientific Acros Supplier No. 315990250
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 13494-80-9 | Te | 127.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13494-80-9
Molecular Formula Te
Molecular Weight (g/mol) 127.60
MDL Number MFCD00134062
InChI Key PORWMNRCUJJQNO-UHFFFAOYSA-N
Synonym tellurium, elemental, telloy, tellur, aurum paradoxum, element, tellur polish, metallum problematum, tellurium, metallic, unii-nqa0o090zj, and compounds
PubChem CID 6327182
ChEBI CHEBI:30452
IUPAC Name tellurium
SMILES [Te]

Specifications

Boiling Point 990.0°C
Melting Point 450.0°C
Color Black to Gray
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.8%
Fieser 15,298
Merck Index 15, 9263
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in h2so4, hno3 and koh, insoluble in benzene and carbon disulfide
Formula Weight 127.6
Percent Purity 99.8%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Tellurium
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.