missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) oxide, 99+%

Product Code. 10042910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10042910 50 g Glass bottle 50g
10793411 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Questo articolo non è restituibile. Consulta la politica di reso
Codice prodotto. 10042910 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 222590500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

CAS: 7446-07-3 | O2Se | 110.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7446-07-3
Molecular Formula O2Se
Molecular Weight (g/mol) 110.97
MDL Number MFCD00003562
InChI Key JPJALAQPGMAKDF-UHFFFAOYSA-N
Synonym selenium oxide, selenium iv oxide, oxoselane oxide, selenious anhydride, selenium oxide seo2, seleniumdioxide, dimer, selenous acid anhydride, selenium iv dioxide 1:2, selenium oxide se2o4
PubChem CID 24007
IUPAC Name (oxo-λ⁴-selanylidene)oxidane
SMILES O=[Se]=O

Specifica

Boiling Point 1245°C
Melting Point 733°C
Color White to Yellow
Physical Form Powder
Quantity 50 g
Assay Percent Range 99+%
Linear Formula TeO2
Merck Index 15, 9265
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in conc. acids and alkalies,slightly soluble in diluted acids
Formula Weight 159.6
Percent Purity ≥99%
Packaging Glass bottle
Density 5.67g/mL
Chemical Name or Material Tellurium(IV) oxide, 99+%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.