missing translation for 'onlineSavingsMsg'
Learn More

Telmisartan Acyl-Beta-D-glucuronide, TRC

Product Code. 30010870
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010870 5 mg 5mg
30134048 2.5 mg 2.5mg
30010032 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010870 Supplier Toronto Research Chemicals Supplier No. TRCT0170105MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 250780-40-6
Molecular Formula C39 H38 N4 O8
Molecular Weight (g/mol) 690.74
Synonym Telmisartan Acyl-B-D-Glucuronide
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[2-[4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propylbenzimidazol-1-yl]methyl]phenyl]benzoyl]oxyoxane-2-carboxylic acid
SMILES CCCc1nc2c(C)cc(cc2n1Cc3ccc(cc3)c4ccccc4C(=O)O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)O)c6nc7ccccc7n6C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C39H38N4O8/c1-4-9-30-41-31-21(2)18-24(36-40-27-12-7-8-13-28(27)42(36)3)19-29(31)43(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)38(49)51-39-34(46)32(44)33(45)35(50-39)37(47)48/h5-8,10-19,32-35,39,44-46H,4,9,20H2,1-3H3,(H,47,48)/t32-,33-,34+,35-,39-/m0/s1
Formula Weight 690.269
Chemical Name or Material Telmisartan Acyl-beta-D-glucuronide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.