missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Telmisartan-d4 (benzyl-2,3,5,6-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Telmisartan-d4 |
| Molecular Formula | C33 D4 H26 N4 O2 |
| Quantity | 0.005 g |
| Synonym | 4’-[(1,4’-Dimethyl-2’-propyl[2,6’-bi-H-benzimidazol]-1’-yl)methyl][1,1’-biphenyl]-2-carboxylic Acid-d4, Micardis-d4, Pritor-d4, 4'-[[4-Methyl-6-(1-methyl-2-benzimidazolyl)-2-propyl-1-benzimidazolyl]methyl]-2-biphenylcarboxylic-d4 Acid |
| InChI Formula | InChI=1 S/C33H30N4O2/c1-4-9-30-35-31-21(2)18-24(32-34-27-12-7-8-13-28(27)36(32)3)19-29(31)37(30)20-22-14-16-23(17-15-22)25-10-5-6-11-26(25)33(38)39/h5-8,10-19 H,4,9,20H2,1-3H3,(H,38,39)/i14D,15 D,16 D,17 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1Cn2c(CCC)nc3c(C)cc(cc23)c4nc5ccccc5n4C)c6ccccc6C(=O)O |
| IUPAC Name | 2-[2,3,5,6-tetradeuterio-4-[[4-methyl-6-(1-methylbenzimidazol-2-yl)-2-propyl-benzimidazol-1-yl]methyl]phenyl]benzoic acid |
| Molecular Weight (g/mol) | 518.64 |
| Formula Weight | 518.262 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction