missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Temozolomide
GREENER_CHOICE

Product Code. 18742744
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18742744 10 mg 10mg
18762694 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18742744 missing translation for 'mfr' Tocris Bioscience™ missing translation for 'supplierNo' 2706/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DNA-methylating antitumor agent

DNA methylating, chemotherapeutic agent. Displays antitumor activity against a board spectrum of tumors, including leukemias, lymphomas and solid tumors (IC50 = 5.0 μM for cytotoxicity against mouse TLX5 lymphoma cells). Induces autophagy in malignant glioma cells. Exhibits enhanced activity when used in combination with CHC (Cat No. 5029).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85622-93-1
Molecular Formula C6H6N6O2

Specifications

Quantity 10 mg
Formula Weight 194.15
Purity >99%
Molecular Weight (g/mol) 194.154
Chemical Name or Material Temozolomide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.