missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ TempDex 710 Nitrile Glove

Product Code. 17668526
Change view
Click to view available options
Size:
7
9
11
Unit Size:
10 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17668526 7 10 pairs
17678526 9 10 pairs
17658526 11 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17668526 Supplier MAPA™ Supplier No. 34710037
  • ONLINE EXCLUSIVE PRICE
    103.75 €
    79.00 € / 10 pairs
    Save 24.75 €
    23% Off
Estimated Shipment: 21-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

TempDex 710 are for high dexterity and thermal protection.

  • Specific advantages
    • Thin, anatomically-shaped glove ensures dexterity
    • Suitable for long-term wear: excellent abrasion resistance
    • Silicone free to prevent traces, defects on metal sheet and glass prior to painting
    • Available in vending machine packaging
  • Specific features
    • Dot embossing for better insulation and better grip
  • Applications
    • Ceramic and plastics industry
      • Handling of composite materials after heat processing
    • Other industries
      • Handling of moulds in the ceramic industry
      • Handling of hot steel after welding
      • Handling of hot piping
    • Construction Industry (Heating engineers/Plumbers)
      • Maintaining pipes, heating equipment

Specifications

Applications Industrial
Color Yellow, Black
Material Nitrile
Size 7
For Use With (Application) Handling of Composite Materials After Heat Processing
Grip Design Dots
Lining Seamless Textile Support
Quantity 50/Cs.
Resistant To Abrasion
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.