missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Temperature Sensor Device for Mastercycler

Product Code. 15753145 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Mastercycler, 384-well
Mastercycler, 96-well
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15753145 Mastercycler, 384-well Each
15733145 Mastercycler, 96-well Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15753145 Supplier Eppendorf™ Supplier No. 0056002006
Estimated Shipment: 18-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid, easy to use, and suitable for use with Mastercycler PCR cyclers.

  • Measuring device without temperature sensor
  • Sensor for 96 well and 384 well blocks, sold separately
  • Simple, rapid verification and adjustment
  • Automatic adjustment with Mastercycler software, version 2.10 or higher
  • Can be third-party adjusted
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Requires Temperature Sensor
For Use With (Equipment) Mastercycler, 384-well
Description Single Channel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.