missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Template Generation System II Kit

Product Code. 10255879 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 reactions
Unit Size:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10255879 20 reactions 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10255879 Supplier Thermo Scientific™ Supplier No. F702
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform transposon-mediated insertion of primer binding sites into target DNA for sequencing or PCR amplification.

Transposon technology is the first technology that allows scientist to insert constructed linear DNA fragment into circular or linear DNA target randomly and very efficiently. The Thermo Scientific Template Generation System II Kit (TGS II Kit) is based on the transposition reaction of the bacteriophage Mu. This system has been simplified to work in vitro, and an artificial Mu transposon, designated as the Entranceposon, has been constructed. The reaction is catalyzed by a single enzyme, MuA Transposase.

TGS II Kit provides the tools for inserting the Entranceposon into foreign DNA at random locations. Insertion of an Entranceposon into unknown target DNA provides primer binding sites for different applications. A simple and fast in vitro transposition reaction takes place in a single reaction tube.

Highlights

  • Provides templates for DNA sequencing more simply and with less hands-on time than any other method
  • Thousands of ready-to-sequence templates from a single transposition reaction: Enough to sequence even large DNA clones (e.g. BAC clones) without fragmentation or subcloning
  • Simple mapping of the Entranceposon insertions by colony-PCR or restriction enzyme digestion enables directed sequencing. Fewer sequencing reactions are required to complete a sequence than with the shotgun approach
  • Universal primers: The sequencing and mapping primers are included in the kit, no need for custom primers.
  • Bidirectional sequencing: A single template clone can be used for sequencing the flanking DNA on both sides of the Entranceposon insertion
  • Three different antibiotic resistance genes are available in each TGS II Kit

All components needed as well as detailed instruction manual to perform the reactions are included in the kit. TGS II Kit is an updated version of the TGS I Kit.

TRUSTED_SUSTAINABILITY

Specifications

Form Solution
Format Tube
Reaction Speed Fast
Product Type Template Generation System II Kit
Quantity 20 reactions
Sufficient For 20 Reactions
For Use With (Application) Pre-amplification
No. of Reactions 20 Reactions
Starting Material DNA

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.