missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tempol
GREENER_CHOICE

Product Code. 18742604
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18742604

Brand: Tocris Bioscience™ 3082/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Superoxide scavenger; antioxidant

Tempol is a superoxide scavenger that displays neuroprotective, anti-inflammatory and analgesic effects. Tempol reduces γ-radiation-induced damage in hematopoietic stem cells in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 2226-96-2
Quantity 100 mg
Molecular Formula C9H18NO2
Formula Weight 172.24
Purity >99%
Chemical Name or Material Tempol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.