missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™ Waterproof Shoulder-Length Cryogenic Gloves

Product Code. 11813821
Change view
Click to view available options
:
Large
Product Code.
11813821 Large
1 options
This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

Product Code. 11813821

Brand: Tempshield™ SHLWP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • Thin, seamless 100% waterproof liner provides protection from spills and splashes
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Certifications/Compliance (Europe) EN 420, EN 511
Description Shoulder Length
Color Blue
Hand Specific
Product Type Waterproof Cryogenic Gloves
Size Large
Temperature (English) Max. -320°F
Temperature (Metric) Max -196°C
EN388 Standard Yes
For Use With (Application) Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes
Series Cryo-Gloves™
Length (English) 24.5 to 27.3 in.
Length (Metric) 62 to 69.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.