Learn More
Thermo Scientific™ Temsirolimus, 99+%
A Rapamycin derivative with anti-cancer activity that inhibits mTOR
285.00 € - 753.00 €
Chemical Identifiers
| CAS | 162635-04-3 |
|---|---|
| Formula Weight | 1030.29g/mol |
| Content And Storage | -30°C to -10°C |
| Format | Powder |
| For Use With (Application) | A Rapamycin derivative with anti-cancer activity that inhibits mTOR |
| Description | White to Off-white |
| Assay | ≥99% |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15445829
|
Thermo Scientific™
J63654.MC |
100 mg |
753.00 €
100mg |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15435829
|
Thermo Scientific™
J63654.MB |
25 mg |
285.00 €
25mg |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15455829
|
Thermo Scientific™
J63654.MF |
50 mg |
506.00 €
50mg |
Estimated Shipment: 03-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 162635-04-3 | |
| -30°C to -10°C | |
| A Rapamycin derivative with anti-cancer activity that inhibits mTOR | |
| ≥99% |
| 1030.29g/mol | |
| Powder | |
| White to Off-white |
Specifications
| White to Off-white | |
| Powder | |
| 162635-04-3 | |
| 1030.29g/mol |
| -30°C to -10°C | |
| A Rapamycin derivative with anti-cancer activity that inhibits mTOR | |
| C56H87NO16 | |
| ≥99% |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.