missing translation for 'onlineSavingsMsg'
Learn More

Tenax™ TA, 20-35 mesh

Product Code. 16574354 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16574354 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16574354 Supplier Thermo Scientific Acros Supplier No. 462230100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 24938-68-9 | C20H18O | 274.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24938-68-9
Melting Point 460.0°C
Molecular Formula C20H18O
Quantity 10 g
InChI Key TWADJGWUKGOPFG-UHFFFAOYSA-N
SMILES COC1=C(C=C(C)C=C1C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 2-methoxy-5-methyl-3-phenyl-1,1'-biphenyl
Molecular Weight (g/mol) 274.36
Chemical Name or Material Tenax* TA
compliance-icons
Product Identifier
  • Tenax TA, 20-35 mesh
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Poly(2,6-diphenyl-1,4-phenylene oxide)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.