missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenidap, Tocris Bioscience™
Description
NSAID that preferentially inhibits COX-1 (IC50 values are < 0.03, 1.2 and > 30 μM for COX-1, COX-2 and 5-lipoxygenase respectively). Inhibits formation of pro-inflammatory arachidonic acid metabolites in isolated human peripheral polymorphonuclear leukocytes. Opener of inward rectifying hKir2.3 channel (EC50 = 402 nM).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 320.75 |
| Percent Purity | >99% |
| Chemical Name or Material | Tenidap |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?